missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ S100A10 Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human S100A10 (NP_002957.1) (Pro 2-Lys 97) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 11144H07E50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15268295

  • £209.25 / 50µg


Estimated Shipment
09-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

S100 calcium binding protein A10 (S100A10), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris, 10% glycerol, pH 8.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
6281
PAGE
50μg
Full Length/Recombinant
S100 Calcium Binding Protein A10
Assay Standard
E. coli
Frozen
12.6kDa
≥90% by SDS-PAGE
Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
ANX2L, ANX2LG, CAL1L, CLP11, Ca[1], GP11, mgC111133, P11, p10
S100A10
Other Proteins
N-His Tag
Videos