missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ RSPO3 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the amino acids (Met 1-Val 146) of human RSPO3 (Q9BXY4-1) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11185H08H1U50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15218345

  • £410.16 / 50µg


Estimated Shipment
05-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

R-Spondin 3 (RSPO3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
84870
PAGE
50μg
C-Terminal
<1 EU/μg
R-spondin 3
Assay Standard
Mammalian
Lyophilized
15.3kDa
>96% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
CRISTIN1, FLJ14440, PWTSR, THSD2
RSPO3
Other Proteins
His Tag
Videos