missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ RSPO1 Recombinant Human Protein, His Tag, 28.2kDa

This recombinant protein was expressed from a DNA sequence encoding the human RSPO1 (NP_001033722.1) (Met 1-Ala 263) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11083H08H5

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15268195

  • £59.86 / 5µg


Estimated Shipment
15-03-2018
Add to basket

Description

Description

R-Spondin 1 (RSPO1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind recombinant mouse CD36 in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
28.2kDa
5μg
Wnt Proteins & Receptors|Other Proteins
RP11-566C13.1, CRISTIN3, FLJ40906, RSPO
RSPO1
Other Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
R-spondin 1
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.