missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ RBP4 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mouse RBP4 (NP_035385.1) (Met 1-Leu 201) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50170M08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15342856

  • £57.26 / 5µg


Estimated Shipment
12-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: One vial of lyophilized protein

Description

Description

Retinol binding protein 4, plasma (RBP4), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind all trans retinoic acid; the binding of retinoic acid results in the quenching of Trp fluorescence in RBP4; the 50% binding concentration (BC50) is >0.5μM
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
22.8kDa
5μg
Other Proteins
Rbp-4, Rbp4
RBP4
Other Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
RBP4 retinol binding protein 4, plasma
Assay Standard
Mammalian
Videos