missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ PVRL2 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Leu 360) of human PVRL2 (NP_002847.1) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10005H08H250

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15274495

  • £676.65 / 250µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Poliovirus receptor-related 2 (herpes virus entry mediator B) (PVRL2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human PVRL2 at 20μg/mL (100μL/well) can bind biotinylated DNAM1 with a linear range of 0.078-2.5μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
36.2kDa
5 x 50μg
Other Proteins|Viral Host Cell Receptors
PVRL2, HVEB, PRR2, CD112, PVRR2
PVRL2
CD Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Poliovirus receptor-related 2
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.