missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ PVR (CD155) Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 345) of mouse PVR precursor (NP_081790.1) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50259M08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13416276

  • £55.70 / 5µg


Estimated Shipment
08-06-2018
Add to basket

Description

Description

Poliovirus receptor (PVR/CD155), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its ability to bind recombinant mouse CD226/DNAM-1; immobilized recombinant mouse PVR at 1μg/mL (100μL/well) can bind recombinant mouse CD226/DNAM-1 with a linear range of 0.78-100ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
52118
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Viral Host Cell Receptors
3830421F03Rik, CD155, D7Ertd458e, HVED, PVS, Taa1, Tage4, mE4
PVR
CD Proteins
His Tag
Lyophilized
28kDa
5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Poliovirus receptor
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.