missing translation for 'onlineSavingsMsg'
Learn More

PVR (CD155) Recombinant Human Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asn 343) of human PVR (NP_006496.3) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Invitrogen 10109H02H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13495706

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Poliovirus receptor (PVR/CD155), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human DNAM1 at 2μg/mL (100μL/well) can bind human PVR-Fc with a linear range of 0.032-0.8μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
61.8kDa
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Poliovirus receptor
IgG1 Fc Tag
CD Proteins
Viral Host Cell Receptors
5817
50μg
Truncated/Recombinant/Cell Surface Protein
PVR, CD155, HVED, NECL5, Necl-5, PVS, TAGE4
PVR
Mammalian
Lyophilized
≥90% by SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.