missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ PVR (CD155) Recombinant Human Protein, DDDDK Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asn 343) of human PVR (NP_006496.3) fused to a DDDDK tag at the N-terminus

Brand:  Thermo Scientific™ 10109HCCH5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13425716

  • £57.00 / 5µg

Estimated Shipment
Add to basket



Poliovirus receptor (PVR/CD155), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human PVR (400ng/well) onto a mouse anti-human PVR antibody coated plate binds recombinant human CD226 with a linear range of 4.0-20μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.



Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Viral Host Cell Receptors
PVR, CD155, HVED, NECL5, Necl-5, PVS, TAGE4
CD Proteins
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Poliovirus receptor
Assay Standard

For Research Use Only. Not for use in diagnostic procedures.