missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ PTX3 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human PTX3 (NP_002843.2) (Met 1-Ser 381) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 12082H08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15285735

  • £82.50 / 5µg


EU Stock 3
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Pentraxin 3, Long (PTX3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5806
PAGE
5μg
C-Terminal
Other Proteins
PTX3, TNFAIP5, TSG-14
PTX3
Other Proteins
His Tag
Lyophilized
41.6kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
<1 EU/μg
Pentraxin 3, long
Assay Standard
Mammalian
Videos