missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ PTPRJ Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human PTPRJ (Q12913) cytoplasmic domain (Arg 997-Ala 1337) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 13165H07E25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15246195

  • £391.42 / 25µg


Estimated Shipment
15-05-2018
Add to basket

Description

Description

Protein Tyrosine Phosphatase, Receptor Type, J (PTPRJ), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5795
PAGE
5 x 5μg
N-Terminal
Tyrosine Phosphatases
CD148, DEP1, HPTPeta, R-PTP-ETA, SCC1
PTPRJ
CD Proteins
His Tag
Lyophilized
41kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
<1 EU/μg
Protein tyrosine phosphatase, receptor type, J
Assay Standard
E. coli
Videos

For Research Use Only. Not for use in diagnostic procedures.