missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ PTPN2 Recombinant Human Protein, N-His.GST Tag

This recombinant protein was expressed from a DNA sequence encoding the amino acid (Met 1-Asn 314) of human PTPN2 (P17706-1) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus

Brand:  Thermo Scientific™ 10570H20B5

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15286945

  • £56.74 / 5µg


Estimated Shipment
16-05-2018
Add to basket

Description

Description

Protein Tyrosine Phosphatase Nonreceptor Type 2 (PTPN2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to dephosphorylate a phosphotyrosine residue in an EGF receptor 988-998 phosphopeptide substrate (R&D Systems Cat. No. ES006), the specific activity is >30nM/min./μg
  • Formulation: Lyophilized in 20mM Tris, 500mM NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
64.4kDa
5μg
Tyrosine Phosphatases
PTN2, PTPT, TC-PTP, TCELLPTP, TCPTP
PTPN2
Other Proteins
GST Tag, His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Protein tyrosine phosphatase, nonreceptor type 2
Assay Standard
Insect
Videos

For Research Use Only. Not for use in diagnostic procedures.