missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ PTPN1 Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human PTPN1 (NP_002818.1) (Glu 2 -Asn 321) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 10304H07E50

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15215135

  • £195.71 / 50µg


Estimated Shipment
02-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Protein tyrosine phosphatase, nonreceptor type 1 (PTPN1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to dephosphorylate a phosphotyrosine residue in an EGF receptor (aa 988-998) phosphopeptide substrate, the specific activity is >15nM/min./μg
  • Formulation: Lyophilized in 10mM HEPES, 500mM NaCl, 1mM DTT, 1mM EDTA, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
38kDa
50μg
Other Proteins
Protein tyrosine phosphatase, nonreceptor type 3
Assay Standard
E. coli
Lyophilized
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
PTP1B, PTPN1
PTPN1
Other Proteins
His Tag
Videos