missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ PTPN1 Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human PTPN1 (NP_002818.1) (Glu 2 -Asn 321) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 10304H07E25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15295125

  • £127.01 / 25µg


Estimated Shipment
08-06-2018
Add to basket

Description

Description

Protein tyrosine phosphatase, nonreceptor type 1 (PTPN1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to dephosphorylate a phosphotyrosine residue in an EGF receptor (aa 988-998) phosphopeptide substrate, the specific activity is >15nM/min./μg
  • Formulation: Lyophilized in 10mM HEPES, 500mM NaCl, 1mM DTT, 1mM EDTA, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
38kDa
5 x 5μg
Other Proteins
Protein tyrosine phosphatase, nonreceptor type 3
Assay Standard
E. coli
Lyophilized
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
PTP1B, PTPN1
PTPN1
Other Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.