missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ PRLR Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human PRLR (NP_000940.1) (Met 1-Asp 234) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10278H08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15225075

  • £57.00 / 5µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Prolactin Receptor Isoform 1 Precursor (PRLR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5618
PAGE
5μg
C-Terminal
<1 EU/μg
Prolactin receptor isoform 1 precursor
Assay Standard
Mammalian
Lyophilized
26kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
hPRLrI, PRLR
PRLR
Cytokines & Receptors
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.