missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ PPIA Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human PPIA (P62937) (Met 1-Glu 165) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10436H08E250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15255405

  • £418.49 / 250µg


Estimated Shipment
10-07-2018
Add to basket

Description

Description

Peptidylprolyl Isomerase A (Cyclophin A) (PPIA), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: 50mM Tris, 150mM NaCl, 20% Glycerol, pH 7.7
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5478
PAGE
5 x 50μg
Full Length/Recombinant
Peptidylprolyl Isomerase A (Cyclophin A)
Assay Standard
E. coli
Frozen
19.4kDa
≥90% by SDS-PAGE
Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
CYPA, CYPH, mgC117158, mgC12404, mgC23397
PPIA
Other Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.