Sino Biological™ PPIA Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human PPIA (P62937) (Met 1-Glu 165) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 10436H08E25

RECOM HU PPIA (HIS)

UNSPSC: 12352202

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15245405

Quantity Price
1 £ 119.0 / 25µg
Estimated Shipment date
from Supplier 08-03-2017
Add to basket


Description and Specification

Specification

Molecular Weight 19.4kDa
Purity ≥90% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Full Length/Recombinant
Gene Alias CYPA, CYPH, mgC117158, mgC12404, mgC23397
Gene Symbol PPIA
Protein Tag His Tag
Validated Application Assay Standard
Expression System E. coli
Format Frozen
Gene ID (Entrez) 5478
Purification Method PAGE
Common Name Peptidylprolyl Isomerase A (Cyclophin A)
Protein Family Other Proteins
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

Peptidylprolyl Isomerase A (Cyclophin A) (PPIA), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: 50mM Tris, 150mM NaCl, 20% Glycerol, pH 7.7
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.