missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ PLAUR (UPAR) Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human UPAR isoform 1 (Q03405-1) (Met 1-Arg 303) without the pro peptide, fused to the polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10925H08H25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15217395

  • £123.88 / 25µg


Estimated Shipment
10-07-2018
Add to basket

Description

Description

Plasminogen Activator, Urokinase Receptor (UPAR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human UPAR at 5μg/μL (100μL/well) can bind biotinylated human UPA with a linear ranger of 40-1000ng/μL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
32.8kDa
5 x 5μg
Other Proteins
CD87, U-PAR, UPAR, URKR
PLAUR
CD Proteins
His Tag
Lyophilized
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Plasminogen activator, Urokinase receptor
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.