Sino Biological™ PLAU Recombinant Human Protein Activated by Trypsin, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human PLAU (NP_002649.1) (Met 1-Leu 431) fused to the polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 10815H08HA5

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.10000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15287225

Quantity Price
1 £ 99.0 / 5µg
Estimated Shipment date
from Supplier 13-03-2017
Add to basket


Description and Specification

Specification

Gene Alias PLAU, ATF, UPA, URK, u-PA
Format Lyophilized
Endotoxin Concentration <1 EU/μg
Gene Symbol PLAU
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Expression System Mammalian
Protein Tag His Tag
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Common Name Plasminogen activator, urokinase
Quantity 5μg
Purity ≥90% by SDS-PAGE
Validated Application Assay Standard
Protein Family Other Proteins
Protein Subtype Proteases
Molecular Weight 18kDa

Plasminogen Activator, Urokinase (PLAU) activated by trypsin, recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • The purified protein was activated by trypsin in vitro
  • Activity: Measured by its ability to cleave a peptide substrate, N-carbobenzyloxy-Gly-Gly-Arg-7-amido-4-methylcoumarin (Z-GGR-AMC), the specific activity is >2000pmole/min./μg
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.