Sino Biological™ PIGR Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 638, 365 Ser/Gly) of human PIGR (NP_002635.2) fused to a C-terminal polyhistidine tag

Overview
Brand: Invitrogen

Manufacturer Part Number: 10131H08H5

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.10000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15214715

Quantity Price
1 £ 71.75 / 5µg
Estimated Shipment date
from Supplier 15-05-2017
Add to basket


Description and Specification

Specification

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene Alias PIGR, FLJ22667, mgC125361, mgC125362
Protein Subtype Fc Receptors
Expression System Mammalian
Molecular Weight 80kDa
Validated Application Assay Standard
Format Lyophilized
Protein Family Other Proteins
Protein Tag His Tag
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Purity ≥90% by SDS-PAGE
Gene Symbol PIGR
Common Name Polymeric immunoglobulin receptor
Endotoxin Concentration <1 EU/μg
Quantity 5μg

Polymeric Immunoglobulin Receptor (PIGR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized human IgM at 2μg/mL (100μL/well) can bind biotinylated PIGR with a linear range of 0.94-15ng/mL; 2. When human IgM is immobilized at 2μg/mL (100μL/well), PIGR inhibits 50% binding of biotinylated PIGR (0.062μg/mL) at the concentration range of 0.03-20μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.