missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ PI3 (ELAFIN) Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human PI3 (NP_002629.1) (Met 1-Gln 117) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 12187H08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15265835

  • £142.62 / 5µg


Estimated Shipment
09-07-2018
Add to basket

Description

Description

Peptidase Inhibitor 3, Skin-Derived (PI3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5266
PAGE
5μg
C-Terminal
<1 EU/μg
Peptidase inhibitor 3, skin-derived
Assay Standard
Mammalian
Lyophilized
11.3kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
ESI, mgC13613, SKALP, WAP3, WFDC14, cementoin
PI3
Proteases, Peptidases & Inhibitors
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.