Sino Biological™ PGF Recombinant Mouse Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the mouse PGF (P49764) (Met 1-Pro 158) fused to the Fc region of human IgG1 at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 50125M02H50

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.
Includes: One vial of protein in suspension buffer

Product Code. 15392766

Quantity Price
1 £ 586.0 / 50µg
EU Stock 1
Add to basket


Description and Specification

Specification

Molecular Weight 42.9kDa
Gene ID (Entrez) 18654
Expression System Mammalian
Validated Application Assay Standard
Format Lyophilized
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Gene Alias AI854365, PIGF, Plgf
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Termination C-Terminal
Protein Family Growth Factors & Receptors
Endotoxin Concentration <1 EU/μg
Gene Symbol PGF
Protein Tag IgG1 Fc Tag
Structural Form Full Length/Recombinant
Common Name Placental growth factor
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Quantity 50μg

Placental Growth Factor (PGF), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.