missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ PECAM1 Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Lys 601) of human PECAM1 (EAW94208.1) fused to the fused Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10148H02H50

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15204745

  • £162.66 / 50µg


Estimated Shipment
12-06-2018
Add to basket

Description

Description

Platelet/Endothelial Cell Adhesion Molecule 1 (PECAM1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by its ability of the immobilized protein to support the adhesion of Jurkat human leukemic T cells; when cells are added to recombinant human PECAM1 coated plates (3.3μg/mL, 100μL/well) in the presence of PMA, approximately 50%-65% will adhere specifically after 30 minutes at 37°C
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
91.2kDa
50μg
Intercellular Adhesion Molecule
PECAM1, CD31, PECAM-1
PECAM1
CD Proteins
IgG1 Fc Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Platelet/endothelial cell adhesion molecule 1
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.