missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ PDPN Recombinant Human Protein, hIgG1-Fc.His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human PDPN short isoform (Q86YL7-1) (Met 1-Lys 123) fused to the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 11065H03H50

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15287695

  • £243.60 / 50µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Podoplanin (PDPN), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human PDPN (Mfr. No. 10012-H08H) at 10μg/mL (100μL/well) can bind biotinylated human CLEC1B-His; the EC50 of biotinylated human CLEC1B-His is 0.71μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
38.6kDa
50μg
Viral Host Cell Receptors
PDPN, CTA-520D8.1, AGGRUS, GP36, GP40, Gp38, HT1A-1, OTS8, PA2.26, T1A, T1A-2
PDPN
Other Proteins
His Tag/IgG1 Fc Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Podoplanin
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.