missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ PDGFRB Recombinant Human Protein (without Catalytic Activity Domain), His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Lys 531) of human PDGFRB (NP_002600.1) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10514H08H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13475886

  • £460.13 / 250µg


Estimated Shipment
08-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Platelet-derived growth factor receptor, beta polypeptide (PDGFRB), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5159
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Receptor Tyrosine Kinases|Growth Factor Receptors
PDGFRB, CD140B, JTK12, PDGFR, PDGFR1
PDGFRB
Growth Factors & Receptors/CD Proteins
PDGFR Kinase Family
Lyophilized
57.6kDa
5 x 50μg
Truncated/Recombinant
<1 EU/μg
Platelet-derived growth factor receptor, beta polypeptide
Assay Standard
Mammalian
His Tag
Videos