missing translation for 'onlineSavingsMsg'
Learn More

PDGFRA Recombinant Human Protein (without Catalytic Activity Domain), His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 524) of human PDGFRA (NP_006197.1) fused to a polyhistidine tag at the C-terminus

Brand:  Invitrogen 10556H08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13435916

  • £271.00 / 50µg

Estimated Shipment date
from Supplier 05-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Platelet-derived growth factor receptor, alpha polypeptide (PDGFRA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human PDGFRA-His at 1μg/well can bind human PDGFC-Fc with a linear range of 0.032-0.8μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5156
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Receptor Tyrosine Kinases|Growth Factor Receptors
PDGFRA, CD140a, mgC74795, PDGFR2, Rhe-PDGFRA
PDGFRA
Growth Factors & Receptors/CD Proteins
PDGFR Kinase Family
Lyophilized
57.7kDa
50μg
Truncated/Recombinant
<1 EU/μg
Platelet-derived growth factor receptor, alpha
Assay Standard
Mammalian
His Tag
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.