missing translation for 'onlineSavingsMsg'
Learn More

PDGFC Recombinant Mouse Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the receptor-binding form of mouse PDGFC (Q8CI19) (Val 235-Gly 345) fused to the Fc region of human IgG1 at the N-terminus

£78.60 - £1685.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias 1110064L01Rik, AI647969, PDGF-C
View More Specs

Includes: Five vials of lyophilized protein

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13433139 View Documents Invitrogen
50458M01H25
5 x 5μg £255.00
50µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 06-02-2020
Please to purchase this item. Need a web account? Register with us today!
13453139 View Documents Invitrogen
50458M01H250
5 x 50μg £1685.00
250µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 06-02-2020
Please to purchase this item. Need a web account? Register with us today!
13443139 View Documents Invitrogen
50458M01H5
5μg £78.60
250µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 06-02-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Platelet-derived growth factor C (PDGFC), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: 1. Measured by its binding ability in a functional ELISA, immobilized mouse PDGFC (100μL/well of a 10μg/mL solution) can bind human PDGFRA; 2. Measured in a cell proliferation assay using BALB/c 3T3 mouse embryonic fibroblast cells, the EC50 for this effect is typically 0.5-2μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
1110064L01Rik, AI647969, PDGF-C

For Research Use Only. Not for use in diagnostic procedures.