missing translation for 'onlineSavingsMsg'
Learn More

PDGFC Recombinant Mouse Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the receptor-binding form of mouse PDGFC (Q8CI19) (Val 235-Gly 345) fused to the Fc region of human IgG1 at the N-terminus

Brand:  Invitrogen 50458M01H5

Code : NEW

Additional Details : Weight : 0.00001kg

Product Code. 13443139

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Includes: One vial of lyophilized protein

Description

Description

Platelet-derived growth factor C (PDGFC), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: 1. Measured by its binding ability in a functional ELISA, immobilized mouse PDGFC (100μL/well of a 10μg/mL solution) can bind human PDGFRA; 2. Measured in a cell proliferation assay using BALB/c 3T3 mouse embryonic fibroblast cells, the EC50 for this effect is typically 0.5-2μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
5μg
Truncated/Recombinant
1110064L01Rik, AI647969, PDGF-C
PDGFC
Mammalian
Lyophilized
≥90% by SDS-PAGE
40.9kDa
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Platelet-derived growth factor, C polypeptide
IgG1 Fc Tag
Growth Factors & Receptors
PDGFs (Platelet-Derived Growth Factors)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.