missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ PDGFC Recombinant Mouse Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the receptor-binding form of mouse PDGFC (Q8CI19) (Val 235-Gly 345) fused to the Fc region of human IgG1 at the N-terminus

Brand:  Thermo Scientific™ 50458M01H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13453139

  • £1651.03 / 250µg


Estimated Shipment
08-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of lyophilized protein

Description

Description

Platelet-derived growth factor C (PDGFC), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: 1. Measured by its binding ability in a functional ELISA, immobilized mouse PDGFC (100μL/well of a 10μg/mL solution) can bind human PDGFRA; 2. Measured in a cell proliferation assay using BALB/c 3T3 mouse embryonic fibroblast cells, the EC50 for this effect is typically 0.5-2μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
40.9kDa
5 x 50μg
Truncated/Recombinant
<1 EU/μg
Platelet-derived growth factor, C polypeptide
Assay Standard
Mammalian
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
PDGFs (Platelet-Derived Growth Factors)
1110064L01Rik, AI647969, PDGF-C
PDGFC
Growth Factors & Receptors
IgG1 Fc Tag
Videos