missing translation for 'onlineSavingsMsg'
Learn More

PDGFC Recombinant Human Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the mature form of human PDGFC (NP_057289.1) (Val 235-Gly 345) fused to the Fc region of human IgG1 at the N-terminus

Brand:  Invitrogen 10273H01H5

Code : NEW

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 13462959

  • £68.23 / 5µg


Estimated Shipment
11-03-2019
Add to basket

Description

Description

Platelet-derived growth factor C (PDGFC), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured in a cell proliferation assay using BALB/c 3T3 mouse embryonic fibroblast cells, the EC50 for this effect is typically 0.5-2μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
39kDa
5μg
Full Length/Recombinant
<1 EU/μg
Platelet derived growth factor C
Assay Standard
Mammalian
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
PDGFs (Platelet-Derived Growth Factors)
FALLOTEIN, SCDGF
PDGFC
Growth Factors & Receptors
IgG1 Fc Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.