missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ PDCD1 Recombinant Mouse Protein, hIgG1-Fc.His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 167) of mouse PDCD1 (NP_032824.1) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 50124M03H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15372756

  • £125.97 / 25µg


Estimated Shipment
08-06-2018
Add to basket

Includes: Five vials of lyophilized protein

Description

Description

Programmed cell death 1 (PDCD1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant mouse CD274 at 1μg/mL (100μL/well) can bind mouse PDCD1 with a linear range of 6.25-400ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
44.2kDa
5 x 5μg
Other Proteins
Pdcd1, Ly101, PD-1, Pdc1
PDCD1
Other Proteins
His Tag/IgG1 Fc Tag
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Programmed cell death 1
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.