missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ PCSK9 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the full length of mouse PCSK9 (NP_705793.1) precursor (Met 1-Gln 694) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 50251M08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15373006

  • £774.51 / 50µg


Estimated Shipment
12-06-2018
Add to basket

Description

Description

Proprotein Convertase Subtilisin/Kexin Type 9 (PCSK9), recombinant mouse protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse PCSK9 at 10μg/mL (100μL/well) can bind biotinylated recombinant human LDLR; the EC50 of biotinylated human LDLR is 0.12μg/mL
  • Formulation: 15mM Tris, 90mM NaCl, 50% glycerol, pH 7.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
72.6kDa
50μg
Proteases
RP23-159J7.1, AI415265, AI747682, FH3, HCHOLA3, mgC47409, Narc1
PCSK9
Mammalian
Frozen
≥90% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Proprotein convertase subtilisin/kexin type 9
Assay Standard
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.