missing translation for 'onlineSavingsMsg'
Learn More

PARP1 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from the amino acids corresponding to the full length of human PARP1 (NP_001609.2) (Met 1-Trp 1014) fused to a polyhistidine tag at the C-terminus

Brand:  Invitrogen 11040H08B25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15227615

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Poly (Adp-Ribose) Polymerase 1 (PARP1), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human PARP1 at 10μg/mL (100μL/well) can bind to biotinylated human HSP70, the EC50 of biotinylated human HSP70 is 0.035μg/mL
  • Formulation: 50mM Tris, 100mM NaCl, pH 8, 2mM EDTA, 2mM PMSF, 0.5mM TCEP, 10% glycerol
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
5 x 5μg
<1 EU/μg
Poly (ADP-ribose) polymerase 1
His Tag
Frozen
≥90% by SDS-PAGE
114.5kDa
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
PARP1, RP11-125A15.2, ADPRT, ADPRT1, PARP, PARP-1, PPOL, pADPRT-1
PARP1
Insect
Other Proteins
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.