missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ PADI4 Recombinant Human Protein, N-His.GST Tag

This recombinant protein was expressed from a DNA sequence encoding the human PADI4 (NP_036519.2) (Met 1-Pro 663) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus

Brand:  Thermo Scientific™ 11072H20B50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15207725

  • £703.72 / 50µg


Estimated Shipment
16-05-2018
Add to basket

Description

Description

Peptidyl Arginine Deiminase, Type IV (PADI4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 20mM Tris, 500mM NaCl, 0.5mM PMSF, 0.5mM TCEP, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
23569
PAGE
50μg
N-Terminal
<1 EU/μg
Peptidyl arginine deiminase, type IV
Assay Standard
Insect
Lyophilized
76kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
PAD, PAD4, PADI5, PDI4, PDI5
PADI4
Other Proteins
GST Tag, His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.