missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ P4HB Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human PH4B (NP_000909.2) corresponding to amino acid (Met 1-Lys 505) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 10827H08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15267255

  • £221.74 / 50µg


Estimated Shipment
06-07-2018
Add to basket

Description

Description

Protein Disulfide-Isomerase (P4HB), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5034
PAGE
50μg
C-Terminal
<1 EU/μg
Prolyl 4-hydroxylase, beta polypeptide
Assay Standard
Mammalian
Lyophilized
60kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
DSI, ERBA2L, GIT, P4Hbeta, PDI, PDIA1, PHDB, PO4DB, PO4HB, PROHB
P4HB
Other Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.