missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ OSMR Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human OSMR (NP_003990.1) extracellular domain (Met 1-Met 740) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11226H08H25

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15278415

  • £123.88 / 25µg


Estimated Shipment
08-06-2018
Add to basket

Description

Description

Oncostatin M Receptor (OSMR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
9180
PAGE
5 x 5μg
C-Terminal
<1 EU/μg
Oncostatin M receptor
Assay Standard
Mammalian
Lyophilized
82.6kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
MGC150626, mgC150627, mgC75127, OSMRB
OSMR
Cytokines & Receptors
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.