missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ NRXN3 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human NRXN3 (NP_620426.2) extracellular domain (Met 1-Thr 357) fused to a polyhistidine tag at the C-terminus

Brand:  Invitrogen 11843H08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15245555

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

Neurexin-3-Beta (NRXN3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of C6 Rat brain glial cells; when 5 x 104 cells/well are added to NRXN3 coated plates (0.8μg/mL and 100μL/well), approximately 30-50% of the added cells will adhere specifically after 60 minutes at 37°C
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
36kDa
5 x 5μg
Other Proteins
C14orf60, KIAA0743, mgC176711
NRXN3
Other Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Neurexin-3-beta
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.