Sino Biological™ NRP2 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Tyr 855) of human NRP2 (NP_003863.2) fused to a C-terminal polyhistidine tag

Overview
Brand: Invitrogen

Manufacturer Part Number: 10695H08H5

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.10000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15277075

Quantity Price
1 £ 80.25 / 5µg
Estimated Shipment date
from Supplier 16-05-2017
Add to basket


Description and Specification

Specification

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Molecular Weight 95kDa
Purity ≥90% by SDS-PAGE
Quantity 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Growth Factor Receptors|Glycoproteins
Endotoxin Concentration <1 EU/μg
Gene Alias NRP2, mgC126574, NP2, NPN2, PRO2714, VEGF165R2
Common Name Neuropilin 2
Gene Symbol NRP2
Validated Application Assay Standard
Protein Family Growth Factors & Receptors
Expression System Mammalian
Protein Tag His Tag

Neuropilin 2 (NRP2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human Neuropilin-2 at 2.5μg/mL (100μL/well) can bind recombinant human NRP2, the EC50 for this effect is typically 0.57nM
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.