missing translation for 'onlineSavingsMsg'
Learn More

NRG4 Recombinant Human Protein, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the human NRG4 (Q8WWG1) extracellular domain (Pro 2-Phe 62) fused to additional two amino acids (Gly & Pro) at the N-terminus

Brand:  Invitrogen 12183HNCE5

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15225835

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Neuregulin 4 (NRG4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 50mM NaCl, pH 7.2, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
6.7kDa
5μg
Truncated/Recombinant
HRG4
NRG4
Growth Factors & Receptors/Cell Structure & Organelle Proteins
EGF (Epidermal Growth Factor)
145957
PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
<1 EU/μg
Neuregulin 1
E. coli
Lyophilized
≥90% by SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.