missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ NRG1 Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human NRG1 isoform alpha (Q02297-1) (Ser 20-Lys 241) fused to the Fc region of human IgG1 at the C-terminus.

Brand:  Thermo Scientific™ 13499H02H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15276315

  • £123.88 / 25µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Neuregulin 1 (NRG1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
51kDa
5 x 5μg
Other Growth Factors
ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, NDF, SMDF
NRG1
Growth Factors & Receptors
IgG1 Fc Tag
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Neuregulin 1
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.