missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ NOV Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the full length of human NOV (NP_002505.1) (Met 1-Met 357) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10264H08B5

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15225045

  • £56.21 / 5µg


Estimated Shipment
13-06-2018
Add to basket

Description

Description

Nephroblastoma Overexpressed (NOV), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of Balb/3T3 mouse embryonic fibroblast cells; when cells are added to NOV coated plates (10μg/mL, 100μL/well), 50%-60% will adhere specifically after 60 minutes at 37°C
  • Formulation: 50mM Tris, 100mM NaCl, 0.5mM PMSF, 10mM Imidazole, 10% glycerol, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
37kDa
5μg
Other Proteins
NOV, CCN3, IGFBP9
NOV
Insect
Frozen
≥90% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Nephroblastoma overexpressed
Assay Standard
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.