missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ NGFR Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the human NGFR (P08138) extracellular domain (Met 1- Asn 250) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 13184H02H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15236215

  • £215.83 / 50µg


Estimated Shipment
07-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Nerve Growth Factor Receptor (NGFR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
4804
PAGE
50μg
C-Terminal
<1 EU/μg
Nerve growth factor receptor
Assay Standard
Mammalian
Lyophilized
50.6kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
CD271, Gp80-LNGFR, TNFRSF16, p75(NTR), p75NTR
NGFR
Growth Factors & Receptors
IgG1 Fc Tag
Videos