missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ NGF Recombinant Mouse Protein

This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse NGF (NP_001106168.1), residues 122 to 241, with an initial Met at the N-terminus

Brand:  Thermo Scientific™ 50385MNAC250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13466286

  • £432.02 / 250µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Nerve growth factor (NGF), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications.
  • Activity: Measured in a cell proliferation assay using TF1 human erythroleukemic cells; the ED50 for this effect is 0.2-2.0ng/mL
  • Formulation: Lyophilized in 50mM Na acetate buffer, pH 5.5, 150mM NaCl, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
18049
≥95% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Neurotrophic Factors
Ngf, Ngfb
NGF
Growth Factors & Receptors
Untagged
Lyophilized
13.5kDa
5 x 50μg
Native
<1 EU/μg
Nerve growth factor
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.