missing translation for 'onlineSavingsMsg'
Learn More

mMP9 Recombinant Mouse Protein, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the mousemMP9 (P41245-1) (Met 1-Pro 730)

Brand:  Invitrogen 50560MNAH25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15348946

  • £475.70 / 25µg


Estimated Shipment
03-04-2019
Add to basket

Includes: Five vials of protein in suspension buffer

Description

Description

Matrix Metallopeptidase 9 (MMP9), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
17395
PAGE
5 x 5μg
Full Length/Recombinant
RP23-61O3.10-001, AW743869, B/MMP9, Clg4b, mMP-9, pro-MMP-9
MMP9
Proteases, Peptidases & Inhibitors
Lyophilized
78.4kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
<1 EU/μg
Matrix metallopeptidase 9
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.