missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ mMP8 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the humanmMP8 (NP_002415.1) (Met 1-Gly 467) fused to the polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10254H08H50

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15215015

  • £774.51 / 50µg


Estimated Shipment
10-05-2018
Add to basket

Description

Description

Matrix metallopeptidase 8 (neutrophil collagenase) (MMP8), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave the fluorogenic peptide substrate, Mca-PLGL-Dpa-AR-NH2, the specific activity is >450pmoles/min./μg
  • Formulation: Lyophilized in 50mM Tris, 10mM CaCl2, 150mM NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
52.6kDa
50μg
Proteases
CLG1, HNC, PMNL-CL, mMP8
MMP8
Other Proteins
His Tag
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Matrix metallopeptidase 8 (neutrophil collagenase)
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.