missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ mMP7 Recombinant Human Protein, N-hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the active form of humanmMP7 (NP_002414.1) (Tyr 95-Lys 267) fused to Fc region of human IgG1 at the N-terminus

Brand:  Thermo Scientific™ 10277H01H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15285065

  • £141.58 / 5µg


Estimated Shipment
08-06-2018
Add to basket

Description

Description

Matrix Metallopeptidase 7 (MMP7), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 100mM Glycine, 10mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
4316
PAGE
5μg
N-Terminal
Proteases
MMP7, mMP-7, MPSL1, PUMP-1
MMP7
Proteases, Peptidases & Inhibitors
IgG1 Fc Tag
Lyophilized
45.8kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
<1 EU/μg
Matrix metallopeptidase 7
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.