missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ MIF Recombinant Mouse Protein

This recombinant protein was expressed from a DNA sequence encoding the mouse MIF (P34884) precursor (Pro 2-Ala 115) fused to an initial Met at the C-terminus

Brand:  Thermo Scientific™ 50066MNAE250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15302636

  • £811.98 / 250µg

Estimated Shipment
Add to basket

Includes: Five vials of protein in suspension buffer



Macrophage Migration Inhibitory Factor (MIF), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 100mM NaCl, pH 8.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.



Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5 x 50μg
Full Length/Recombinant
GIF, Glif, mgC107654
Cytokines & Receptors
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
<1 EU/μg
Macrophage migration inhibitory factor
Assay Standard
E. coli

For Research Use Only. Not for use in diagnostic procedures.