Sino Biological™ MIF Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human MIF (P14174) (Pro 2-Ala 115) fused to a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 10246H08H50

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15235005

Quantity Price
1 £ 508.0 / 50µg
EU Stock 3
Add to basket


Description and Specification

Specification

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 4282
Molecular Weight 13.8kDa
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination C-Terminal
Structural Form Full Length/Recombinant
Endotoxin Concentration <1 EU/μg
Gene Alias GIF, GLIF, mMIF
Common Name Macrophage migration inhibitory factor
Gene Symbol MIF
Validated Application Assay Standard
Protein Family Cytokines & Receptors
Expression System Mammalian
Protein Tag His Tag

Macrophage Migration Inhibitory Factor (MIF), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.