missing translation for 'onlineSavingsMsg'
Learn More

MET (c-MET) Recombinant Human Protein (without Catalytic Activity Domain), His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 932) of human MET (NP_000236) fused to a polyhistidine tag at the C-terminus

£74.50 - £722.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias MET, AUTS9, HGFR, RCCP2, c-Met
Gene ID (Entrez) 4233
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13445926 View Documents Invitrogen
10692H08H5
5μg £74.50
5µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 15-04-2020
Please to purchase this item. Need a web account? Register with us today!
13455926 View Documents Invitrogen
10692H08H50
50μg £241.00
50µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 29-04-2020
Please to purchase this item. Need a web account? Register with us today!
13435926 View Documents Invitrogen
10692H08H250
5 x 50μg £722.00
250µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 15-04-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Met proto-oncogene/hepatocyte growth factor receptor (MET/c-MET), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human HGF at 10μg/mL (100μL/well) can bind biotinylated MET, the ED50 of biotinylated Met is 5.28μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
4233
<1 EU/μg
MET, AUTS9, HGFR, RCCP2, c-Met

For Research Use Only. Not for use in diagnostic procedures.