missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ MET (c-MET) Recombinant Human Protein (without Catalytic Activity Domain), His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 932) of human MET (NP_000236) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10692H08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13455926

  • £237.35 / 50µg


Estimated Shipment
29-05-2018
Add to basket

Description

Description

Met proto-oncogene/hepatocyte growth factor receptor (MET/c-MET), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human HGF at 10μg/mL (100μL/well) can bind biotinylated MET, the ED50 of biotinylated Met is 5.28μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
4233
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Receptor Tyrosine Kinases|Growth Factor Receptors
MET, AUTS9, HGFR, RCCP2, c-Met
MET
Growth Factors & Receptors
MET (HGFR) Kinase Family
Lyophilized
103.0kDa
50μg
Truncated/Recombinant
<1 EU/μg
Met proto-oncogene
Assay Standard
Mammalian
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.