missing translation for 'onlineSavingsMsg'
Learn More

MET (c-MET) Recombinant Human Protein (without Catalytic Activity Domain), His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 932) of human MET (NP_000236) fused to a polyhistidine tag at the C-terminus

Brand:  Invitrogen 10692H08H250

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 13435926

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Met proto-oncogene/hepatocyte growth factor receptor (MET/c-MET), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human HGF at 10μg/mL (100μL/well) can bind biotinylated MET, the ED50 of biotinylated Met is 5.28μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
103.0kDa
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Met proto-oncogene
MET (HGFR) Kinase Family
Mammalian
Lyophilized
≥85% by SDS-PAGE
4233
5 x 50μg
Truncated/Recombinant
MET, AUTS9, HGFR, RCCP2, c-Met
MET
His Tag
Growth Factors & Receptors
Receptor Tyrosine Kinases, Growth Factor Receptors
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.